CutMaster Para-Aramid Flame Resistant Sleeves with Thumb Strap, Magid
Supplier: Magid Glove
Certificates
About this item
Magid® CutMaster machine knit 100% para-aramid protective sleeves with elastic thumb strap offer exceptional cut, puncture, abrasion, heat and flame resistance.
- 100% para-aramid machine knit, seamless, standard width, two-ply construction
- Available in a 3" diameter in lengths from 9 to 21"
- Exceptional cut, puncture, abrasion, heat and flame resistance
- Overcast edge
- ANSI cut level 4
CutMaster knit para-aramid sleeves may be washed with no loss of their protective properties and with minimal shrinkage. Each yellow seamless sleeve features an overcast edge to help prevent unraveling.
Protection sleeves come in the right length to allow for a snug and comfortable fit that minimizes obstruction while working. Elastic thumb strap on these sleeve styles ensure they stay in place while allowing excellent agility and dexterity.
Certifications: ANSI ASTM F-1790-97 Testing Standards.
Specifications
- Catalog No:
- 11000-270
- Garment application:
- Industrial, Fire protection, Laboratory
- Risk area:
- Flame-retardant