Order Entry
ContactUsLinkComponent
Nalgene® Unitary™ Color-Coded Wash Bottles, Low-Density Polyethylene, Thermo Scientific
Nalgene® Unitary™ Color-Coded Wash Bottles, Low-Density Polyethylene, Thermo Scientific
Catalog #: 16125-170
Color-Coded Unitary* Wash Bottles, LDPE
Nalgene® Unitary™ Color-Coded Wash Bottles, Low-Density Polyethylene, Thermo Scientific
Catalog #: 16125-170

Specifications

  • Capacity:
    500 ml (16.9 oz.)
  • Cat. no.:
    16125-170
  • Supplier No.:
    2423-0500

Specifications

About this item

Covenient, pre-packaged Unitary™ wash bottles come with four polypropylene color closures (one each of white, red, yellow, and blue) for easy identification.

Ordering information: A sample of PolyPaper Right-To-Know Custom Labeling System with Mylar® overlay is included in each pack.
Customers Who Bought This Also Bought