Heat-Resistant Terrycloth Pads, Wells Lamont
Supplier: Wells Lamont
Certificates
About this item
Pads provide cut and heat resistance in food processing or industrial applications.
- Protect against up to 325 °F
- Ambidextrous
- Extra heavyweight
- Full thumbs
- Open tops
Customers Who Bought This Also Bought