Service for pH Meters
Supplier: Mettler Toledo
New Product
Certificates
About this item
Professional installation completed by a trained Mettler Toledo service technician will guarantee maximum performance and optimal use.
- Achieve peak performance and compliance with quality systems and regulatory requirements
- Take the right steps at the beginning of equipment use
IPac includes: Documented proof of proper installation (IQ) and taking-to-operation (OQ), user training, and routine tests defined.
EQPac includes: Fully compliant qualification documentation and meter monitoring logbook, and the Standard Calibration Certificate (SCC) designed to evaluate the performance of the pH meter against factory specifications. All calibration data Is recorded and displayed with a Pass/Fail Indication of each parameter.
Customers Who Bought This Also Bought