Perchloric acid 70%, AR Select® ACS, Macron Fine Chemicals™
Catalog #: CAMK882814
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 7601-90-3
MOC: Non-RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, clear, Mall
- Size:500 ml
- Cat. no.:CAMK882814
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- ACS - Assay (HClO₄):69.0 - 72.0 %
- ACS - Chloride (Cl):≤ 0.001 %
- ACS - Color (APHA):≤ 10
- ACS - Heavy Metals (as Pb):≤ 0.0001 %
- ACS - Nitrogen Compounds (as N):≤ 0.001 %
- ACS - Residue after Ignition:≤ 0.003 %
- ACS - Silicate and Phosphate (as SiO₂):≤ 0.0005 %
- ACS - Sulfate (SO₄):≤ 0.001 %
- Determine by Flame Photometry & ICP - Aluminum (Al):≤ 100 ppb
- Determine by Flame Photometry & ICP - Barium (Ba):≤ 100 ppb
- Determine by Flame Photometry & ICP - Beryllium (Be):≤ 10 ppb
- Determine by Flame Photometry & ICP - Bismuth (Bi):≤ 10 ppb
- Determine by Flame Photometry & ICP - Boron (B):≤ 1000 ppb
- Determine by Flame Photometry & ICP - Cadmium (Cd):≤ 50 ppb
- Determine by Flame Photometry & ICP - Calcium (Ca):≤ 1000 ppb
- Determine by Flame Photometry & ICP - Chromium (Cr):≤ 100 ppb
- Determine by Flame Photometry & ICP - Cobalt (Co):≤ 50 ppb
- Determine by Flame Photometry & ICP - Copper (Cu):≤ 100 ppb
- Determine by Flame Photometry & ICP - Gallium (Ga):≤ 500 ppb
- Determine by Flame Photometry & ICP - Germanium (Ge):≤ 500 ppb
- Determine by Flame Photometry & ICP - Gold (Au):≤ 500 ppb
- Determine by Flame Photometry & ICP - Iron (Fe):≤ 100 ppb
- Determine by Flame Photometry & ICP - Lead (Pb):≤ 100 ppb
- Determine by Flame Photometry & ICP - Lithium (Li):≤ 500 ppb
- Determine by Flame Photometry & ICP - Magnesium (Mg):≤ 100 ppb
- Determine by Flame Photometry & ICP - Manganese (Mn):≤ 50 ppb
- Determine by Flame Photometry & ICP - Mercury (Hg):≤ 100 ppb
- Determine by Flame Photometry & ICP - Molybdenum (Mo):≤ 10 ppb
- Determine by Flame Photometry & ICP - Nickel (Ni):≤ 100 ppb
- Determine by Flame Photometry & ICP - Potassium (K):≤ 500 ppb
- Determine by Flame Photometry & ICP - Silicon (Si):≤ 500 ppb
- Determine by Flame Photometry & ICP - Silver (Ag):≤ 100 ppb
- Determine by Flame Photometry & ICP - Sodium (Na):≤ 1000 ppb
- Determine by Flame Photometry & ICP - Strontium (Sr):≤ 100 ppb
- Determine by Flame Photometry & ICP - Thallium (Tl):≤ 50 ppb
- Determine by Flame Photometry & ICP - Tin (Sn):≤ 500 ppb
- Determine by Flame Photometry & ICP - Vanadium (V):≤ 10 ppb
- Determine by Flame Photometry & ICP - Zinc (Zn):≤ 100 ppb
- Determine by Flame Photometry & ICP - Zirconium (Zr):≤ 10 ppb
- Determine by Flame Photometry & ICP:
- Appearance (clear, colorless liquid):
- Macron Fine Chemicals™, Perchloric acid 70%, AR Select® ACS:
Specification Test Results
About this item
Boiling Pt:160…181 °C (1013 hPa)
Melting Pt:–112.2 °C
Density:1.53…1.768 g/cm³ (20 °C)
ADR:5.1,I
:Length:
:Width:
:Height:
:Ø
:Thickness:
:70%
:70
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt: 160…181 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –112.2 °C
:Flash Pt:
:Flash Pt:
:M.W. N/A N/A
EINECS:231-512-4
:160…181 °C (1013 hPa)
UN:1873
:5.1
:I
:GHS03
:GHS05
:GHS07
Formula:HClO₄
MW:100.46 g/mol
Storage Temperature:Ambient
:M.O.C. Category N
MDL Number:MFCD00011325
CAS Number:7601-90-3
Customers Who Bought This Also Bought