Phenolphthalein 1.0% in 95% alcohol, StandARd®, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
Formula:C₂₀H₁₄O₄
MW:318.33 g/mol
Boiling Pt:∼78 °C (1013 hPa)
Density:∼0.85 g/cm³ (20 °C)
MDL Number:MFCD00005913
CAS Number:77-09-8
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt: ∼78 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 258 to 262 °C
:Flash Pt:
:Flash Pt:
:M.W. 318.33 g/mol
:Phenolphthalein
EINECS:201-004-7
:∼78 °C (1013 hPa)
UN:1170
:3
:II
:GHS02
:GHS07
:GHS08
Storage Temperature:Ambient
:M.O.C. Category N
:12,07392
:14,07243
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- pH:≥8.0
- Product Information (not specifications): Appearance (clear, colorless:
- solution):
- in 95% alcohol:
Specification Test Results
Related Information
Certificates
Search by Lot NumberAvantor Portfolio of Chemicals
Product Literature
H306 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
sds_MACRH306
SDS
Customers Who Bought This Also Bought