Potassium Chloride, 0.0100 Molar (M/100), 1412 µS/cm (1412 µmho/cm) at 25 °C, Ricca Chemical Company
Supplier: Ricca Chemical
Certificates
About this item
APHA for conductance.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.