BIOXP® 9600 NGS Kits for Plasmids
Supplier: TELESIS BIO
New Product
Certificates
About this item
Generate Illumina-compatible, adapter-ligated, NGS libraries for plasmid sequence confirmation in a simple automated workflow using the BioXp 9600.
- Input: 10 to 20 ng of purified plasmid
- Compatible with Illumina sequencers
- 15 minutes of hands on time
- Compatible with BioXp 9600
The BioXp NGS library prep kit for plasmid sequencing provides the necessary reagents, when combined with customer-provided sequencing adapters, to generate Illumina-compatible NGS libraries for confirmation of full plasmid sequences. This kit uses a streamlined, fully automated ligation-based library prep method. The input into the kit is 10 to 20 ng of purified plasmids in nuclease-free water, Tris-HCl pH 8.0, or Tris low EDTA (0.1 mM).
Delivery: Not supplied with necessary sequencing adapters.
Related Information
BioXp® 9600 NGS Kit for Plasmid Sequencing - Loading Map and Checklist
Additional Product Details
Telesis Bio BioXp® NGS Library Prep Kits
Product Literature
Customers Who Bought This Also Bought