VWR® EUV/ES/PUV/PS Ultrapure Water Systems
Supplier: VWR International
Certificates
About this item
Our VWR® laboratory-grade water purification systems feature an inspiring, application-oriented design. Perform your workflows faster and more reliably, simplifying your daily lab work while ensuring cost-efficient operation over the long term. All instruments offer flexible solutions as they can be adapted to your requirements and integrated into your laboratory environment.
- Delivers consistently high-water quality for reliable and reproducible results; optionally available with UV lamp
- Easy to operate using the touch-activated color display; with direct access to all important dispensing options
- Depending on the model type, with unique Bagtank technology to save you from the hassle of time-consuming tank cleaning
- Space-saving footprint with a width of only 28 cm to easily fit in any lab environment
VWR® Ultrapure laboratory water systems consistently deliver the highest quality water. The compact VWR® Ultrapure laboratory water systems have been designed for Type 1 ultrapure water requirements of 10 liters per day and are ideal for use in the preparation of buffers, media, and samples, both in life sciences applications and in analytical laboratory procedures.
BodyTextAccessories: VWR® Ultrapure UV lamp - Horizontal installation, optimized temperature gradient. Effectively destroys organic compounds. Easy replacement.
VWR® Ultrapure pre-treatment cartridge - Fast and effective adsorption of impurities by high-grade activated carbon. Highly efficient catalyzer for removing oxidating agents such as chlorine. Highly efficient reverse osmosis membranes; optimized water usage. Low-energy membranes for ecological and economic operation.
VWR® Ultrapure polishing cartridge - High performance capacity due to efficient ion exchange resins. Fast and effective adsorption of impurities by high-grade activated carbon. Optimized flow prevents separation of the mixed-bed resin. Patented connection method; easy exchange of consumables.
VWR® Ultrapure sterile filter - Excellent service life and flow rates. Easy to install. Automatic venting. Certified quality.
VWR® Ultrapure storage bag - Fast and easy replacement of the bag. High user safety as the Bagtank eliminates the need for cleaning chemicals.
VWR® Ultrapure Ultra filter - Effective removal of RNase|Dnase. Reliable removal of endotoxins. High flow rate performance. Sterile-packaged.
VWR® Ultrapure pre-treatment cartridge - Fast and effective adsorption of impurities by high-grade activated carbon. Highly efficient catalyzer for removing oxidating agents such as chlorine. Highly efficient reverse osmosis membranes; optimized water usage. Low-energy membranes for ecological and economic operation.
VWR® Ultrapure polishing cartridge - High performance capacity due to efficient ion exchange resins. Fast and effective adsorption of impurities by high-grade activated carbon. Optimized flow prevents separation of the mixed-bed resin. Patented connection method; easy exchange of consumables.
VWR® Ultrapure sterile filter - Excellent service life and flow rates. Easy to install. Automatic venting. Certified quality.
VWR® Ultrapure storage bag - Fast and easy replacement of the bag. High user safety as the Bagtank eliminates the need for cleaning chemicals.
VWR® Ultrapure Ultra filter - Effective removal of RNase|Dnase. Reliable removal of endotoxins. High flow rate performance. Sterile-packaged.
Ordering information: VWR® Ultrapure EUV water system (76777-142), UV lamp included – MUST order the following required consumables: S-Pack 76777-144, Final Filter 76777-126.
VWR® Ultrapure ES water system (76777-140) – MUST order the following required consumables: S-Pack 76777-144, Final Filter 76777-126.
VWR® Ultrapure PUV water system (76777-138), UV lamp included – MUST order the following required consumables: S-Pack 76777-136, Final Filter 76777-126, Storage Bag 76777-128, Pre-treatment Filter 76777-132.
VWR® Ultrapure PS water system (76777-136) – MUST order the following required consumables: S-Pack 76777-136, Final Filter 76777-126, Storage Bag 76777-128, Pre-treatment Filter 76777-132.
VWR® Ultrapure ES water system (76777-140) – MUST order the following required consumables: S-Pack 76777-144, Final Filter 76777-126.
VWR® Ultrapure PUV water system (76777-138), UV lamp included – MUST order the following required consumables: S-Pack 76777-136, Final Filter 76777-126, Storage Bag 76777-128, Pre-treatment Filter 76777-132.
VWR® Ultrapure PS water system (76777-136) – MUST order the following required consumables: S-Pack 76777-136, Final Filter 76777-126, Storage Bag 76777-128, Pre-treatment Filter 76777-132.
Delivery: Delivered without purification pack, sterile filter and ultrafilter (all sold separately).
Specifications
- Catalog No:
- 76777-128
- 76777-126
- 76777-132
- 76777-130
- 76777-134
- 76777-138
- 76777-136
- 76777-142
- 76777-140
- 76777-144
- Model:
- Ultrapure PUV
- Ultrapure PS
- Ultrapure EUV
- Ultrapure ES
- Average daily needs:
- <10 L
- <10 L
- <10 L
- <10 L
- Max. operating pressure:
- 6.9 bar
- 6.9 bar
- 6.9 bar
- 6.9 bar
- Inorganics @ 25°C:
- 0.055 μS/cm
- 0.055 μS/cm
- 0.055 μS/cm
- 0.055 μS/cm
- Bacteria:
- <0.01 CFU/ml
- <0.01 CFU/ml
- <0.01 CFU/ml
- <0.01 CFU/ml
- Endotoxin:
- <0.001 EU/ml
- <0.001 EU/ml
- <0.001 EU/ml
- <0.001 EU/ml
- DNases:
- <5 pg/ml
- <5 pg/ml
- <5 pg/ml
- <5 pg/ml
- W×D×H:
- 280×530.7×509.4 mm
- 280×530.7×509.4 mm
- 280×530.7×509.4 mm
- 280×530.7×509.4 mm
Specifications
Customers Who Bought This Also Bought