Amyl acetate mixture of isomers, purified, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
Synonyms: n-Pentyl acetate,Acetic acid amyl ester,Acetic acid pentyl ester
Formula:C₇H₁₄O₂
MW:130.19 g/mol
Boiling Pt:149 °C (1013 hPa)
Melting Pt:–71 °C
Density:0.876 g/cm³ (20 °C)
Flash Pt:23 °C (closed cup)
MDL Number:MFCD00009500
CAS Number:628-63-7
ADR:3,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt: 149 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –71 °C
:Flash Pt:
:Flash Pt: 23 °C (closed cup)
:M.W. 130.19 g/mol
:Amyl acetate
EINECS:211-047-3
UN:1104
:3
:III
:GHS02
Storage Temperature:Ambient
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Identification (by IR):Passes Test
- J.T.Baker®, Amyl acetate mixture of isomers, purified:
Specification Test Results
Related Information
Certificates
Search by Lot Number9094 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Customers Who Bought This Also Bought