Tin(II) chloride dihydrate 98.0-103.0%, crystals, AR® ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
Synonyms: Stannous chloride dihydrate
Formula:SnCl₂·2H₂O
MW:225.65 g/mol
Boiling Pt:623 °C (1013 hPa)
Melting Pt:246 °C
Density:2.71 g/cm³ (25 °C)
MDL Number:MFCD00149863
CAS Number:10025-69-1
ADR:8,III
Merck Index:12,08939
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,0-103,0
:Boiling Pt:
:Boiling Pt: 623 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 246 °C
:Flash Pt:
:Flash Pt:
:M.W. 225.65 g/mol
:Tin(II) chloride dihydrate
EINECS:231-868-0
UN:3260
:8
:III
:GHS05
:GHS07
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- ACS - Assay (SnCl₂·2H₂O):98.0 - 103.0 %
- ACS - Mercury (Hg):≤0.050 ppm
- ACS - Solubility in HCl:Passes Test
- ACS - Sodium (Na):≤0.01 %
- ACS - Potassium (K):≤0.005 %
- ACS - Iron (Fe):≤0.003 %
- ACS - Calcium (Ca):≤0.005 %
- ACS - Lead (Pb):≤0.01 %
- Appearance: colorless to slightly yellow (but definitely not green) crystals:
- Macron Fine Chemicals™, Tin(II) chloride dihydrate 98.0-103.0%, crystals, AR® ACS:
Specification Test Results
Related Information
Certificates
Search by Lot Number8176 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
sds_MACR8176
SDS
Customers Who Bought This Also Bought