Toluene ≥99.7% (by GC), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog #: CAMK448310
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 108-88-3
MOC: Non-RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, amber, narrow mouth
- Size:4 L
- Cat. no.:CAMK448310
- Supplier No.:4483-10
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Assay (C₆H₅CH₃) (by GC):≥99.7 %
- Color (APHA):≤10
- Residue after Evaporation:≤4 ppm
- Substances Darkened by H₂SO₄:Passes Test
- Sulfur Compounds (as S):≤0.003 %
- Water (H₂O)(by KF, coulometric):≤0.03 %
- Optical Absorbance (1-cm path vs water)::Passes Test
- Absorbance - 285 nm:≤1.000
- Absorbance - 286 nm:≤0.700
- Absorbance - 288 nm:≤0.400
- Absorbance - 293 nm:≤0.200
- Absorbance - 300 nm:≤0.104
- Absorbance - 310 nm:≤0.050
- Absorbance - 335 nm:≤0.020
- Absorbance 350 - 400 nm:≤0.010
Specification Test Results
About this item
Synonyms: Methylbenzene
Boiling Pt:110.6 °C (1013 hPa)
Melting Pt:–95 °C
Density:0.867 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,7
:Management of Change (MOC) category = N
Flash Pt:4 °C
:Boiling Pt:
:Boiling Pt: 110.6 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –95 °C
:Flash Pt:
:Flash Pt: 4 °C
:M.W. 92.14 g/mol
Merck Index:12,09667
Formula:C₆H₅CH₃
MW:92.14 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00008512
CAS Number:108-88-3
EINECS:203-625-9
UN:1294
REACH:05-2114615130-69
:3
:II
:GHS02
:GHS08
:GHS07
:M.O.C. Category N
Customers Who Bought This Also Bought