L(+)-Tartaric acid 99.7-100.5% (dried basis), fine-granular NF, FCC, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
Synonyms: L-Tartaric Acid,(2R,3R)-(+)-Tartaric acid,Tartaric Acid,(R,R)-(+)-2,3-Dihydroxysuccinic acid,(R,R)-(+)-Tartaric acid,(2R,3R)-2
Formula:C₄H₆O₆
MW:150.09 g/mol
Boiling Pt:399 °C (1013 hPa)
Melting Pt:206 °C
Density:1.759 g/cm³ (20 °C)
Flash Pt:> 100 °C (closed cup)
MDL Number:MFCD00064207
CAS Number:87-69-4
Merck Index:12,09237
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,7-100,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 399 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 206 °C
:Flash Pt:
:Flash Pt: > 100 °C (closed cup)
:M.W. 150.09 g/mol
Storage Temperature:Ambient
EINECS:201-766-0
REACH:01-2119537204-47
:> 100 °C
:GHS05
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- GMP Manufactured Product:
- Meets N.F. Requirements:
- Meets F.C.C. Requirements:
- Food GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- Bulk Food Chemical:
- NF - Assay (HOCO(CHOH)₂COOH) (dried basis)(by acidimetry):99.7 - 100.5 %
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Loss on Drying:≤0.5 %
- NF - Residue on Ignition:≤0.1 %
- NF - Limit of Oxalate:Passes Test
- NF - Sulfate (SO₄):Passes Test
- NF - Optical Rotation [Å]²⁵^D (+):12.2 - 13.0 Degree
- FCC - Assay (HOCO(CHOH)₂COOH) (dried basis)(by acidimetry):99.7 - 100.5 %
- FCC - Identification A (IR):Passes Test
- FCC - Identification B (Tartrate):Passes Test
- FCC - Lead (Pb):≤2 mg/kg
- FCC - Limit of Oxalate:Passes Test
- FCC - Loss on Drying:≤0.5 %
- FCC - Residue on Ignition:≤0.05 %
- FCC - Specific (Optical) Rotation [Å]²⁵^D (+):12.2 - 13.0 Degree
- FCC - Sulfate (SO₄):Passes Test
- Solubility (20 in 100):Passes Test
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
Specification Test Results
Related Information
Certificates
Search by Lot Number2307 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
sds_MACR2307
SDS
Customers Who Bought This Also Bought