Triethylamine ≥99.5%, BAKER BIO-ANALYZED®, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: (Diethylamino)ethane
Formula:(C₂H₅)₃N
MW:101.19 g/mol
Boiling Pt:90 °C (1013 hPa)
Melting Pt:–115 °C
Density:0.729 g/cm³ (20 °C)
Flash Pt:–11 °C
MDL Number:MFCD00009051
CAS Number:121-44-8
ADR:3,II
Merck Index:12,09799
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt: 90 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –115 °C
:Flash Pt:
:Flash Pt: –11 °C
:M.W. N/A N/A
:Triethylamine
Storage Temperature:Ambient
EINECS:204-469-4
UN:1296
:3
:II
:–11 °C
:GHS02
:GHS05
:GHS07
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Assay ((CH₃CH₂)₃N):≥ 99.0 %
- Color (APHA):≤ 15
- Identification (by IR):Passes Test
- Water (H₂O):≤ 1 %
Specification Test Results
Related Information
Certificates
Search by Lot Number9111 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
spec_JT9111-7
Product specification
Customers Who Bought This Also Bought