ActivArmr® 07-111 / 07-112 Vibration Reducing Gloves, Ansell
Supplier: Ansell Healthcare
Certificates
About this item
ActivArmr® gloves are specifically engineered to reduce the amount of vibration absorbed by the hands when using power tools and other vibrating machinery.
- Padding reduces vibrations
- Superior worker protection
- High abrasion and cut resistance
- All gloves feature a Velcro® closure
Gloves are suitable for operating riveting, grinding, sanding, and polishing tools. Soft Gelfom™ padding helps reduce vibration and protect workers from Hand-Arm Vibration Syndrome (HAVS). In addition, a nitrile coating provides snag, puncture, abrasion, and cut resistance, while also helping to keep hands warm and dry.
Half-finger style has coated palms, canvas back, and padding that extends from the palm to the end of each half-finger. Full-finger style is fully coated with padding that extends from the palm to the end of each finger.
Ordering information: Gloves are available in pairs and as single right- or left-handed gloves.
Specifications
- Catalog No:
- 80092-648
- 80092-646
- 80092-630
- 80092-650
- 80092-644
- 80092-622
- 80092-654
- Model:
- 07-112
- 07-112
- 07-112
- 07-112
- 07-112
- 07-111
- 07-112
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Cotton/foam
- Cotton/foam
- Cotton/foam
- Cotton/foam
- Cotton/foam
- Cotton/foam
- Cotton/foam
- Finish:
- Fully impregnated
- Fully impregnated
- Fully impregnated
- Fully impregnated
- Fully impregnated
- Impregnated palm
- Fully impregnated
- Cuff style:
- Hook & loop / Velcro® fastening
- Hook & loop / Velcro® fastening
- Hook & loop / Velcro® fastening
- Hook & loop / Velcro® fastening
- Hook & loop / Velcro® fastening
- Hook & loop / Velcro® fastening
- Hook & loop / Velcro® fastening
- Anti-static:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes