2-Propanol ≥99.5%, CMOS for microelectronic, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: IPA,Isopropyl Rubbing Alcohol,Isopropyl alcohol,Dimethyl carbinol,2-Hydroxy propane,Isopropanol
Formula:(CH₃)₂CHOH
MW:60.1 g/mol
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.786 g/cm³ (20 °C)
Flash Pt:12 °C (closed cup)
MDL Number:MFCD00011674
CAS Number:67-63-0
ADR:3,II
Merck Index:13,05228
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 82 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -89 °C|Melting Pt: –89 °C
:Flash Pt:
:Flash Pt: 12 °C (closed cup)
:M.W. 60.1 g/mol
:2-Propanol
EINECS:200-661-7
UN:1219
REACH:01-2119457558-25
:3
:II
:GHS02
:GHS07
Storage Temperature:Ambient
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Microelectronic Use:
- Assay (CH₃CHOHCH₃):≥ 99.5 %
- Color (APHA):≤ 10
- Residue after Evaporation:≤ 4 ppm
- Solubility in H₂O:Passes Test
- Water (H₂O)(by Karl Fischer titrn):≤ 0.05 %
- Acidity (µeq/g):≤ 0.2
- Alkalinity (µeq/g):≤ 0.1
- Chloride (Cl):≤ 0.2 ppm
- Phosphate (PO₄):≤ 0.3 ppm
- Trace Impurities - Aluminum (Al):≤ 50.0 ppb
- Arsenic and Antimony (as As):≤ 10.0 ppb
- Trace Impurities - Barium (Ba):≤ 20.0 ppb
- Trace Impurities - Beryllium (Be):≤ 100.0 ppb
- Trace Impurities - Bismuth (Bi):≤ 100.0 ppb
- Trace Impurities - Boron (B):≤ 10.0 ppb
- Trace Impurities - Cadmium (Cd):≤ 20.0 ppb
- Trace Impurities - Calcium (Ca):≤ 50.0 ppb
- Trace Impurities - Chromium (Cr):≤ 20.0 ppb
- Trace Impurities - Cobalt (Co):≤ 20 ppb
- Trace Impurities - Copper (Cu):≤ 10.0 ppb
- Trace Impurities - Gallium (Ga):≤ 50 ppb
- Trace Impurities - Germanium (Ge):≤ 50.0 ppb
- Trace Impurities - Gold (Au):≤ 20 ppb
- Heavy Metals (as Pb):≤ 200.0 ppb
- Trace Impurities - Iron (Fe):≤ 50.0 ppb
- Trace Impurities - Lead (Pb):≤ 20.0 ppb
- Trace Impurities - Lithium (Li):≤ 50.0 ppb
- Trace Impurities - Magnesium (Mg):≤ 20 ppb
- Trace Impurities - Manganese (Mn):≤ 15.0 ppb
- Trace Impurities - Molybdenum (Mo):≤ 100.0 ppb
- Trace Impurities - Nickel (Ni):≤ 10.0 ppb
- Trace Impurities - Niobium (Nb):≤ 100.0 ppb
- Trace Impurities - Potassium (K):≤ 100 ppb
- Trace Impurities - Silicon (Si):≤ 50 ppb
- Trace Impurities - Silver (Ag):≤ 20.0 ppb
- Trace Impurities - Sodium (Na):≤ 100.0 ppb
- Trace Impurities - Strontium (Sr):≤ 20.0 ppb
- Trace Impurities - Tantalum (Ta):≤ 100.0 ppb
- Trace Impurities - Thallium (Tl):≤ 10.0 ppb
- Trace Impurities - Tin (Sn):≤ 100 ppb
- Trace Impurities - Titanium (Ti):≤ 20 ppb
- Trace Impurities - Vanadium (V):≤ 100.0 ppb
- Trace Impurities - Zinc (Zn):≤ 50 ppb
- Trace Impurities - Zirconium (Zr):≤ 100.0 ppb
- Particle Count at point of fill - 0.5 µm and greater (Rion KS42AF):≤ 150 par/ml
- Particle Count at point of fill - 1.0 µm and greater (Rion KS42AF):≤ 25 par/ml
Specification Test Results
Related Information
Certificates
Search by Lot Number9059 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
spec_JT9059-4S spec_JT9059-4X spec_JT9059-9
Product specification
Customers Who Bought This Also Bought