L(+)-Tartaric acid ≥99.5%, granular NF, Multi-Compendial, J.T.Baker®
Catalog #: CAJT4105-5
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 87-69-4
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Poly bottle, white, blue T/E cap
- Size:2.5 kg
- Supplier No.:4105-05
Specifications
Specification Test Results
- Meets E.P. Chemical Specifications:
- Meets B.P. Chemical Specifications:
- Meets J.P. Chemical Specifications:
- GMP Manufactured Product:
- Meets N.F. Requirements:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Specific Rotation [Å]²⁵^D(calculated on dried basis)(+):12.0 - 13.0 Degree
- NF - Loss on Drying:≤0.5 %
- NF - Residue on Ignition:≤0.1 %
- NF - Oxalate (C₂O₄):Passes Test
- NF - Sulfate (SO₄):Passes Test
- NF - Assay (HOCO(CHOH)₂COOH) (dried basis):99.7 - 100.5 %
- NF - Endotoxin Concentration (EU/g):≤10
- EP/BP - Assay (HOCO(CHOH)₂COOH) (dried basis):99.5 - 101.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Specific Rotation [Å]²º^D (+):12.0 - 12.8 Degree
- EP/BP - Oxalic Acid:≤360 ppm
- EP/BP - Chloride (Cl):≤100 ppm
- EP/BP - Sulfate (SO₄):≤150 ppm
- EP/BP - Calcium (Ca):≤200 ppm
- EP/BP - Loss on Drying at 105°C:≤0.2 %
- EP/BP - Ash (sulfated):≤0.1 %
- JP - Assay (HOCO(CHOH)₂COOH) (dried basis):99.7 - 101.0 %
- JP - Identification A:Passes Test
- JP - Identification B:Passes Test
- JP - Sulfate (SO₄):≤0.048 %
- JP - Oxalate (C₂O₄):Passes Test
- JP - Heavy Metals (as Pb):≤10 ppm
- JP - Calcium (Ca):Passes Test
- JP - Arsenic (As):≤1 ppm
- JP - Loss on Drying:≤0.5 %
- JP - Residue on Ignition:≤0.05 %
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- The USP Organic Volatile Impurities <467> requirements for this product is met by the implementation of Residual Solvents <467> as per the USP Revision Bulletin of June 15, 2007.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
Specification Test Results
About this item
Synonyms: L-Tartaric Acid,(2R,3R)-(+)-Tartaric acid,Tartaric Acid,(R,R)-(+)-2,3-Dihydroxysuccinic acid,(R,R)-(+)-Tartaric acid,(2R,3R)-2
Boiling Pt:399 °C (1013 hPa)
Melting Pt:168…170 °C
Density:1.759 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
:Management of Change (MOC) category = R
Flash Pt:150 °C (closed cup)
:Boiling Pt:
:Boiling Pt: 399 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 168 to 170 °C|Melting Pt: 168…170 °C
:Flash Pt:
:Flash Pt: 150 °C (closed cup)
:M.W. 150.09 g/mol
Merck Index:12,09237
Formula:C₄H₆O₆
MW:150.09 g/mol
Storage Temperature:Ambient
EINECS:201-766-0
REACH:01-2119537204-47
:150 °C
:GHS05
:M.O.C. Category R
MDL Number:MFCD00064207
CAS Number:87-69-4
Customers Who Bought This Also Bought