Spec-Tec® Spectra® Fiber Sterile Critical Environment Glove Liners, Wells Lamont
Supplier: Wells Lamont
Certificates
About this item
Cut-resistant liners are ideal for pathologists, coroners, hospital waste handlers, and orthopedic, trauma, and general surgeons.
- Cut-resistant liners feature excellent tactile sensitivity
- Spectra fiber for moderate cut resistance with high reusability
- Knit cuff
- White
- Ambidextrous and sterile
These ambidextrous, sterile liners feature excellent tactile sensitivity.
Spectra fiber is an ultrahigh molecular weight polyethylene fiber that provides moderate cut resistance with high reusability.
Specifications
- Catalog No:
- 80091-126
- 80091-124
- 80091-122
- 76302-800
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Sterility:
- Sterile
- Sterile
- Sterile
- Sterile