Sodium thiosulfate pentahydrate 99.5-101.0%, crystals, AR®, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Formula:Na₂S₂O₃.5H₂O
MW:248.19 g/mol
Melting Pt:48 °C
Density:1.67 g/cm³ (20 °C)
MDL Number:MFCD00149186
CAS Number:10102-17-7
Merck Index:12,08844
:Sodium thiosulfate pentahydrate
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5-101,0
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 48 °C
:Flash Pt:
:Flash Pt:
:M.W. 248.19 g/mol
Storage Temperature:Ambient
EINECS:231-867-5
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Assay (Na₂S₂O₃·5H₂O):99.5 - 101.0 %
- Appearance (colorless, transparent crystals):Passes Test
- Insoluble Matter:≤0.005 %
- Nitrogen Compounds (as N):≤0.002 %
- pH of 5% Solution at 25°C:6.0 - 8.4
- Solubility:Passes Test
- Sulfate and Sulfite (as SO₄):≤0.1 %
- Sulfide (S):Passes Test
- Water (H₂O)(by Karl Fischer titrn), For Information Only:
- Macron Fine Chemicals™, Sodium thiosulfate pentahydrate 99.5-101.0%, crystals, AR®:
Specification Test Results
Related Information
Certificates
Search by Lot Number8100 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
sds_MACR8100
SDS
Customers Who Bought This Also Bought