tri-Sodium citrate dihydrate 99.0-100.5% (by anhydrous basis), crystals, GenAR® USP, Multi-Compendial, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: Sodium Citrate Dihydrate,Sodium citrate tribasic dihydrate,Sodium Citrate, Dihydrate
Formula:Na₃C₆H₅O₇·2H₂O
MW:294.1 g/mol
Melting Pt:150 °C
Density:1.76 g/cm³ (20 °C)
MDL Number:MFCD00150031
CAS Number:6132-04-3
Merck Index:13,08675
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-100,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 150 °C
:Flash Pt:
:Flash Pt:
:M.W. 294.1 g/mol
:tri-Sodium citrate dihydrate
Storage Temperature:Ambient
EINECS:200-675-3
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- GMP Manufactured Product:
- Meets B.P. Chemical Specifications:
- Meets J.P. Chemical Specifications:
- Meets E.P. Chemical Specifications:
- Meets U.S.P Requirements:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Assay (C₆H₅Na₃O₇) (anhydrous basis):99.0 - 100.5 %
- USP - Alkalinity:Passes Test
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Identification C:Passes Test
- USP - Tartrate:Passes Test
- USP - Water (H₂O):10.0 - 13.0 %
- EP/BP - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis):99.0 - 101.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Acidity or Alkalinity:Passes Test
- EP/BP - Chloride (Cl):≤50 ppm
- EP/BP - Oxalate (C₂O₄):≤300 ppm
- EP/BP - Sulfate (SO₄):≤150 ppm
- EP/BP - Readily Carbonizable Substances:Passes Test
- EP/BP - Water (H₂O):11.0 - 13.0 %
- EP/BP - Appearance of Solution:Passes Test
- Endotoxin Concentration (EU/g):≤5
- JP - Assay (anhydrous basis):99.0 - 101.0 %
- JP - pH (1 in 20):7.5 - 8.5
- JP - Clarity and Color of Solution:Passes Test
- JP - Chloride (Cl):≤0.015 %
- JP - Identification:Passes Test
- JP - Sulfate (SO₄):≤0.048 %
- JP - Heavy Metals (as Pb):≤10 ppm
- JP - Arsenic (As):≤2 ppm
- JP - Tartrate:Passes Test
- JP - Oxalate (C₂O₄):Passes Test
- JP - Readily Carbonizable Substances:Passes Test
- JP - Loss on Drying at 180°C:10.0 - 13.0 %
- Appearance (fine, white to slightly yellow crystals):
- No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.:
- Macron Fine Chemicals™, tri-Sodium citrate dihydrate 99.0-100.5% (by anhydrous basis), crystals, GenAR® USP, Multi-Compendial:
Specification Test Results
Related Information
Certificates
Search by Lot Number7773 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Macron 7773 Sodium Citrate Dihydrate
Datasheet
sds_BAKR7773
SDS
Customers Who Bought This Also Bought