Sodium hydroxide ≥98% (by acidimetry), pellets, AR® ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
New Product
MOC: Non-Regulated
About this item
Synonyms: Caustic soda
Formula:NaOH
MW:40 g/mol
Boiling Pt:1390 °C (1013 hPa)
Melting Pt:323 °C
Density:2.13 g/cm³ (20 °C)
MDL Number:MFCD00003548
CAS Number:1310-73-2
ADR:8,II
Merck Index:14,08627
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt: 1390 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 323 °C
:Flash Pt:
:Flash Pt:
:M.W. 40 g/mol
EINECS:215-185-5
UN:1823
REACH:01-2119457892-27
:8
:II
:GHS05
Storage Temperature:Ambient
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- Assay (NaOH) (by acidimetry):≥98 %
- Identification:Passes Test
- Calcium (Ca):≤0.005 %
- Chloride (Cl):≤0.005 %
- Copper (Cu):≤0.001 %
- Heavy Metals (as Ag):≤0.001 %
- Insoluble Matter:≤0.003 %
- Iron (Fe):≤0.0003 %
- ACS - Magnesium (Mg):≤0.002 %
- Mercury (Hg):≤0.1 ppm
- Nickel (Ni):≤0.0005 %
- Nitrogen Compounds (as N):≤0.0003 %
- Phosphate (PO₄):≤0.0002 %
- Potassium (K):≤0.02 %
- Sodium Carbonate (Na₂CO₃):≤0.4 %
- ACS - Sulfate (SO₄):≤0.003 %
- Meets Reagent Specifications for testing USP/NF monographs:
- Appearance (white hygroscopic pellets):
- Macron Fine Chemicals™, Sodium hydroxide ≥98% (by acidimetry), pellets, AR® ACS:
Specification Test Results
Related Information
Certificates
Search by Lot Number7708 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Macron 7708.pdf
Datasheet
spec_MK770806 spec_MK770810 spec_MK770812 spec_MK770820
Product specification
Customers Who Bought This Also Bought