Sodium chloride 99.0-100.5% (dried basis), granular USP, FCC, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
Synonyms: Saline,Sea salt
Formula:NaCl
MW:58.44 g/mol
Boiling Pt:1413 °C (1013 hPa)
Melting Pt:801 °C
Density:2.16 g/cm³ (20 °C)
MDL Number:MFCD00003477
CAS Number:7647-14-5
Merck Index:14,08599
:Sodium chloride
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-100,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 1413 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 801 °C
:Flash Pt:
:Flash Pt:
:M.W. 58.44 g/mol
EINECS:231-598-3
:GHS07
Storage Temperature:Ambient
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets F.C.C. Requirements:
- Food GMP Manufactured Product:
- GMP Manufactured Product:
- Meets U.S.P Requirements:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- Bulk Food Chemical:
- USP - Assay (NaCl)(USP) (dried basis):99.0 - 100.5 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Loss on Drying at 105°C:≤0.5 %
- USP - Acidity or Alkalinity:Passes Test
- USP - Iodide (I):Passes Test
- USP - Barium (Ba):Passes Test
- USP - Magnesium and Alkaline Earth Metals (as Ca):≤100 ppm
- USP - Arsenic (As):≤1 ppm
- USP - Nitrite (NO₂) (Absorbance Value):≤0.01
- USP - Sulfate (SO₄):≤200 ppm
- USP - Phosphate (PO₄):≤25 ppm
- USP - Iron (Fe):≤2 ppm
- USP - Bromide (Br):≤100 ppm
- USP - Appearance of Solution:Passes Test
- USP - Ferrocyanide:Passes Test
- USP - Aluminum (Al):≤0.2 ppm
- USP - Potassium (K):≤500 ppm
- FCC - Assay (as NaCl) (FCC) (ignited basis):99.0 - 100.5 %
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- FCC - Arsenic (As):≤1 ppm
- FCC - Calcium and Magnesium:≤0.35 %
- FCC - Heavy Metals (as Pb):≤2 ppm
- FCC - Loss on Drying at 105°C:≤0.5 %
- Solubility (10 in 50):Passes Test
- Appearance (white, crystalline granules, slightly hygroscopic):
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Must be subjected to further processing during the preparation of:
- injectable dosage forms.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- Macron Fine Chemicals™, Sodium chloride 99.0-100.5% (dried basis), granular USP, FCC:
Specification Test Results
Related Information
Certificates
Search by Lot Number7532 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals VWR Top Rated Guide
Product Literature
Macron 7532 Sodium Chloride
Datasheet
sds_MACR7532
SDS
spec_MK753206
Product specification
Customers Who Bought This Also Bought