Sodium hydrogen sulfate monohydrate, white crystals, AR®, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: Sodium bisulphate monohydrate
Formula:NaHSO₄·H₂O
MW:138.08 g/mol
Melting Pt:183 °C
Density:2.1 g/cm³ (20 °C)
MDL Number:MFCD00149210
CAS Number:10034-88-5
ADR:8,III
Merck Index:13,08658
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 183 °C
:Flash Pt:
:Flash Pt:
:M.W. 138.08 g/mol
EINECS:231-665-7
:183
UN:3260
:8
:III
:GHS05
Storage Temperature:Ambient
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Solubility (10 in 50):Passes Test
- Chloride (Cl):≤0.001 %
- Acidity (as H₂SO₄):35 - 37 %
- Arsenic (As):≤0.0001 %
- Insoluble Matter and NH₄OH Precipitate:≤0.005 %
- Calcium and Magnesium Precipitate:≤0.005 %
- Heavy Metals (as Pb):≤0.0005 %
- Trace Impurities - Iron (Fe):≤0.001 %
- Phosphate (PO₄):≤0.001 %
- Appearance (white crystals):
- Macron Fine Chemicals™, Sodium hydrogen sulfate monohydrate, white crystals, AR®:
Specification Test Results
Related Information
Certificates
Search by Lot Number7432 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
sds_MACR7432
SDS
Customers Who Bought This Also Bought