Sulfuric acid 95.0-98.0% (by acidimetry), AR Select® ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
Formula:H₂SO₄
MW:98.08 g/mol
Boiling Pt:∼335 °C (1013 hPa)
Melting Pt:3 °C
Density:1.84 g/cm³ (20 °C)
MDL Number:MFCD00064589
CAS Number:7664-93-9
Merck Index:14,08974
:98.08
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95,0-98,0
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt: ∼335 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 3 °C
:Flash Pt:
:Flash Pt:
:Density: 1,84 g/cm³ (20 °C)
:M.W. 98.08 g/mol
EINECS:231-639-5
:∼335 °C (1013 hPa)
UN:1830
:8
:II
:1.84
:GHS05
:GHS07
:GHS08
Storage Temperature:Ambient
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- ACS - Assay (H₂SO₄) (by acidimetry):95.0 - 98.0 %
- Appearance:Passes Test
- ACS - Ammonium (NH₄):≤ 0.5 ppm
- ACS - Arsenic and Antimony (as As):≤ 0.003 ppm
- ACS - Chloride (Cl):≤ 0.05 ppm
- ACS - Color (APHA):≤ 8
- ACS - Heavy Metals (as Pb):≤ 0.1 ppm
- ACS - Nitrate (NO₃):≤ 0.1 ppm
- ACS - Residue after Ignition:≤ 1 ppm
- ACS - Substances Reducing Permanganate (as SO₂):≤ 2 ppm
- Determine by Flame Photometry & ICP - Aluminum (Al):≤ 50.0 ppb
- Determine by Flame Photometry & ICP - Barium (Ba):≤ 10.0 ppb
- Determine by Flame Photometry & ICP - Beryllium (Be):≤ 50.0 ppb
- Determine by Flame Photometry & ICP - Bismuth (Bi):≤ 100.0 ppb
- Determine by Flame Photometry & ICP - Boron (B):≤ 10.0 ppb
- Determine by Flame Photometry & ICP - Cadmium (Cd):≤ 5.0 ppb
- Determine by Flame Photometry & ICP - Calcium (Ca):≤ 50.0 ppb
- Determine by Flame Photometry & ICP - Chromium (Cr):≤ 10.0 ppb
- Determine by Flame Photometry & ICP - Cobalt (Co):≤ 5.0 ppb
- Determine by Flame Photometry & ICP - Copper (Cu):≤ 5.0 ppb
- Determine by Flame Photometry & ICP - Gallium (Ga):≤ 100.0 ppb
- Determine by Flame Photometry & ICP - Germanium (Ge):≤ 100.0 ppb
- Determine by Flame Photometry & ICP - Gold (Au):≤ 100.0 ppb
- Determine by Flame Photometry & ICP - Iron (Fe):≤ 50.0 ppb
- Determine by Flame Photometry & ICP - Lead (Pb):≤ 1.0 ppb
- Determine by Flame Photometry & ICP - Lithium (Li):≤ 10.0 ppb
- Determine by Flame Photometry & ICP - Magnesium (Mg):≤ 50.0 ppb
- Determine by Flame Photometry & ICP - Manganese (Mn):≤ 5.0 ppb
- Determine by Flame Photometry & ICP - Mercury (Hg):≤ 1.0 ppb
- Determine by Flame Photometry & ICP - Molybdenum (Mo):≤ 10 ppb
- Determine by Flame Photometry & ICP - Nickel (Ni):≤ 5.0 ppb
- Determine by Flame Photometry & ICP - Potassium (K):≤ 50.0 ppb
- Determine by Flame Photometry & ICP - Silicon (Si):≤ 100.0 ppb
- Determine by Flame Photometry & ICP - Silver (Ag):≤ 10.0 ppb
- Determine by Flame Photometry & ICP - Sodium (Na):≤ 500.0 ppb
- Determine by Flame Photometry & ICP - Strontium (Sr):≤ 10.0 ppb
- Determine by Flame Photometry & ICP - Thallium (Tl):≤ 200.0 ppb
- Determine by Flame Photometry & ICP - Tin (Sn):≤ 50.0 ppb
- Determine by Flame Photometry & ICP - Vanadium (V):≤ 10 ppb
- Determine by Flame Photometry & ICP - Zinc (Zn):≤ 5.0 ppb
- Determine by Flame Photometry & ICP - Zirconium (Zr):≤ 10 ppb
- Determine by Flame Photometry & ICP:
- Macron Fine Chemicals™, Sulfuric acid 95.0-98.0% (by acidimetry), AR Select® ACS:
Specification Test Results
Related Information
Certificates
Search by Lot Number5557 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Customers Who Bought This Also Bought