One-Size-Fits-All Nylon Glove Liners, Wells Lamont
Supplier: Wells Lamont
Certificates
About this item
Knitted construction provides a one-size-fits-all option to help reduce on-hand glove inventories.
Knitted construction provides a one-size-fits-all option to help reduce on-hand glove inventories. Stretch nylon has limited reuse. Ambidextrous. Available in half-finger and full-finger styles. Full-finger liner may also be used as a gowning or inspection glove.
- Knitted construction
- Ambidextrous
- Half-finger and full-finger styles available
- Full-finger liner also works as gowning or inspection glove
Stretch nylon has limited reuse.
The full-finger liner may also be used as a gowning or inspection glove.
The ambidextrous gloves are available in half-finger and full-finger styles.
Ordering information: Packaged in poly bags of 12 pair.
Specifications
- Catalog No:
- 32935-536
- 32935-480
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous