Reusable Nylon Glove Liners, Wells Lamont
Supplier: Wells Lamont
Certificates
About this item
Reusable liners are constructed of high-stretch nylon for maximum comfort and superior tactile sensitivity.
- Ambidextrous
- Washable
- Full-finger and half-finger styles available
- Other applications: gowning or inspection gloves
- High-stretch nylon construction
They provide a physical barrier to protect hands from the irritating effects of overexposure to latex, vinyl, and nitrile thin-film gloves.
The ambidextrous and washable gloves are available in full-finger and half-finger styles.
They can also be used as gowning or inspection gloves.
Ordering information: Packaged in poly bags of 12 pair.
Specifications
- Catalog No:
- 32935-561
- 32935-563
- 32935-559
- 32935-569
- 32935-565
- 32935-567
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous