Light Task Plus II™ Gloves, Polyurethane-Coated Palms, Honeywell Safety
Supplier: Honeywell Safety Products
Certificates
About this item
Gloves provide protection in dry conditions while handling materials such as glass, ceramics, and abrasive items such as blocks and lumber.
- Uncoated area on the back of the glove provides ventilation and comfort
- Knit wrist prevents materials from getting inside the glove
Comfortable, seamless Dyneema® fiber liner and palm coated with polyurethane offer excellent abrasion and tear resistance as well as reliable cut protection.
Specifications
- Catalog No:
- CA82030-812
- CA82030-810
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Cuff style:
- Knit wrist
- Knit wrist