Sodium hydroxide 0.05 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated, With Information
About this item
Synonyms: Caustic soda
Formula:NaOH
MW:40 g/mol
Density:1 g/cm³ (25 °C)
MDL Number:MFCD00003548
CAS Number:1310-73-2
ADR:8,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = NI
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 323 °C
:Flash Pt:
:Flash Pt:
:M.W. 40 g/mol
Storage Temperature:Ambient
EINECS:215-185-5
:323
:1390
UN:1824
:8
:III
:M.O.C. Category NI
:12,08772
:14,08627
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Protect from air to avoid absorption of carbon dioxide.:
- Normality:0.0498 - 0.0502
- Chloride (Cl):≤5 ppm
- ACS - Heavy Metals (as Pb):≤1 ppm
- Trace Impurities - Iron (Fe):≤0.5 ppm
- Standardization at 25°C traceable to NIST Standard Reference Material.:
- SRM No…:
Specification Test Results
Related Information
Certificates
Search by Lot Number5664 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Brand Definition Baker Analyzed
Datasheet
Customers Who Bought This Also Bought