Vikan® Heavy-Duty Push Broom Heads
Supplier: Remco Products
New Product
Certificates
About this item
Heavy-duty push brooms feature densely packed stiff and soft bristles to sweep both fine and larger debris.
- Soft/stiff bristle design
- Exposed bristle length: 2.4"
- Polypropylene, polyester, stainless steel construction
- Available in multiple colors to support color-coded area and task segregation plans
Designed from US food and beverage manufacturer feedback. The broom head is heavier, making it easiser to maintain contact with the floor during sweeping. The angled cut of the bristles also helps prevent skipping or hopping during long pushes across production flooring. Flared bristles reach into corners and wall junctions to make cleaning easier and more thorough.
Customers Who Bought This Also Bought