di-Potassium hydrogen phosphate ≥98.0% (dried basis) FCC, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated Limited
About this item
Synonyms: di-Potassium hydrogen orthophosphate,Potassium hydrogen phosphate,Dipotassium hydrogen phosphate,Dibasic Potassium Phosphate,Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate)
Formula:K₂HPO₄
MW:174.18 g/mol
Melting Pt:340 °C
Density:2.3 g/cm³ (20 °C)
MDL Number:MFCD00011383
CAS Number:7758-11-4
Merck Index:12,07828
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,0
:Management of Change (MOC) category = RL
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 340 °C
:Flash Pt:
:Flash Pt:
:M.W. 174.18 g/mol
Storage Temperature:Ambient
EINECS:231-834-5
:M.O.C. Category RL
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets F.C.C. Requirements:
- Food GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Food Chemical:
- FCC - Identification:Passes Test
- FCC - Assay (K₂HPO₄) (dried basis):≥98.0 %
- FCC - Insoluble Substances:≤0.2 %
- FCC - Loss on Drying:≤2.0 %
- FCC - Arsenic (As):≤3 ppm
- FCC - Fluoride (F):≤10 ppm
- FCC - Lead (Pb):≤2 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number3254 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
sds_BAKR3254
SDS
spec_JT3254-1
Product specification
Customers Who Bought This Also Bought