Sulfuric acid ≥96% VLSI, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
Formula:H₂SO₄
MW:98.08 g/mol
Boiling Pt:∼335 °C (1013 hPa)
Melting Pt:3 °C
Density:1.84 g/cm³ (20 °C)
MDL Number:MFCD00064589
CAS Number:7664-93-9
Merck Index:14,08974
:98.08
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:96
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: ∼335 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 10.38 °C|Melting Pt: 3 °C
:Flash Pt:
:Flash Pt:
:Density: 1,84 g/cm³ (20 °C)
:M.W. 98.08 g/mol
EINECS:231-639-5
:∼335 °C (1013 hPa)
UN:1830
:8
:II
:1.84
:GHS05
Storage Temperature:Ambient
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Microelectronic Use:
- Recommended Storage Conditions: 15° - 100°F:
- Assay (H₂SO₄):95.0 - 97.0 %
- Color (APHA):≤10
- Chloride (Cl):≤0.1 ppm
- Nitrate (NO₃):≤0.2 ppm
- Phosphate (PO₄):≤0.3 ppm
- Trace Impurities - Aluminum (Al):≤50.0 ppb
- Arsenic and Antimony (as As):≤5.0 ppb
- Trace Impurities - Barium (Ba):≤50.0 ppb
- Trace Impurities - Beryllium (Be):≤10.0 ppb
- Trace Impurities - Bismuth (Bi):≤20.0 ppb
- Trace Impurities - Boron (B):≤10.0 ppb
- Trace Impurities - Cadmium (Cd):≤50 ppb
- Trace Impurities - Calcium (Ca):≤100.0 ppb
- Trace Impurities - Chromium (Cr):≤50 ppb
- Trace Impurities - Cobalt (Co):≤50.0 ppb
- Trace Impurities - Copper (Cu):≤10.0 ppb
- Trace Impurities - Gallium (Ga):≤20.0 ppb
- Trace Impurities - Germanium (Ge):≤100.0 ppb
- Trace Impurities - Gold (Au):≤40.0 ppb
- Trace Impurities - Iron (Fe):≤200.0 ppb
- Trace Impurities - Lead (Pb):≤20.0 ppb
- Trace Impurities - Lithium (Li):≤50.0 ppb
- Trace Impurities - Magnesium (Mg):≤50.0 ppb
- Trace Impurities - Manganese (Mn):≤10.0 ppb
- Trace Impurities - Molybdenum (Mo):≤10.0 ppb
- Trace Impurities - Nickel (Ni):≤50 ppb
- Trace Impurities - Niobium (Nb):≤10.0 ppb
- Trace Impurities - Potassium (K):≤100 ppb
- Trace Impurities - Silicon (Si):≤100.0 ppb
- Trace Impurities - Silver (Ag):≤50 ppb
- Trace Impurities - Sodium (Na):≤100.0 ppb
- Trace Impurities - Strontium (Sr):≤10.0 ppb
- Trace Impurities - Tantalum (Ta):≤10.0 ppb
- Trace Impurities - Thallium (Tl):≤20.0 ppb
- Trace Impurities - Tin (Sn):≤50 ppb
- Trace Impurities - Titanium (Ti):≤10.0 ppb
- Trace Impurities - Vanadium (V):≤10.0 ppb
- Trace Impurities - Zinc (Zn):≤50 ppb
- Trace Impurities - Zirconium (Zr):≤10.0 ppb
- Particle Count - 0.5 µm and greater:≤80 par/ml
- Particle Count - 1.0 µm and greater:≤10 par/ml
- Reported value is the average of all samples counted for this lot number,with no individual sample value exceeding the specification.:
- J.T.Baker®, Sulfuric acid ≥96% VLSI:
Specification Test Results
Related Information
Certificates
Search by Lot Number5374 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
sds_BAKR5374
SDS
spec_JT5374-3
Product specification
Customers Who Bought This Also Bought