VWR® Bagged Examination Gloves, Nitrile
Supplier: VWR International
New Product
Certificates
About this item
VWR® bagged examination gloves provide quality and comfort. Perfect for cleanroom, laboratory, and ESD environments.
- Bagged gloves for cleanroom compatibility
- Powder-free nitrile
- Cleanroom class 10,000 (DI washed)
- Static control
- Surface resistance: 10¹¹ Ω
- Volume resistance: 10⁹ Ω
VWR® bagged examination gloves are powder-free nitrile, cleanroom compatable gloves that offer static control and provide a high level of quality and comfort. These gloves are available in a blue or white finish.
Certifications: CAFE RGP: 10⁹ ohms as per ANSI/ESD SP15.1.
Packaging: Gloves are packaged 100 per double bag, 10 bags per case (1000 gloves per case).
Specifications
- Catalog No:
- 77257-060
- 77257-058
- 77257-064
- 77257-070
- 77257-068
- 77257-066
- 77257-072
- 77257-062
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cleanroom class:
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- Material:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Anti-static:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes