Thermo Scientific™ Nunc™ Multiwell Plates with Nunclon™ Supra Surface Treatment and Lid
Supplier: Thermo Fisher Scientific
New Product
Certificates
About this item
Experience optimal performance with Thermo Scientific Nunc™ multiwell plates with the Nunclon™ supra surface treatment for culturing a wide variety of fastidious adherent cells. Intended for general cell culture applications, the Nunclon™ supra surface is more hydrophilic than the Nunclon™ delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
- Consistent lot-to-lot performance
- Sterile with SAL 10⁻⁶
- Non-cytotoxic
- Non-pyrogenic
- Biocompatible per USP Class VI for raw materials
- Shelf life five years
Nunclon™ supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Thus, Nunclon™ supra cultureware is a great xeno-free solution for cell therapy research.
Packaging: Individually wrapped.
Customers Who Bought This Also Bought