Potassium hydroxide ≥85.0%, pellets, BAKER ANALYZED®, Electronic Grade, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: Caustic potash
Formula:KOH
MW:56.11 g/mol
Boiling Pt:1320 °C (1013 hPa)
Melting Pt:360 °C
Density:2.04 g/cm³ (20 °C)
MDL Number:MFCD00003553
CAS Number:1310-58-3
ADR:8,II
Merck Index:12,07806
:Length:
:Width:
:Height:
:Ø
:Thickness:
:85,0
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 1320 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 360 °C
:Flash Pt:
:Flash Pt:
:M.W. 56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:1813
REACH:01-2119487136-33
:8
:II
:GHS05
:GHS07
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Microelectronic Use:
- Assay (KOH):≥85.0 %
- Potassium Carbonate (K₂CO₃):≤1.0 %
- Insoluble Matter:≤50 ppm
- Chloride (Cl):≤0.01 %
- Sulfate (SO₄):≤20 ppm
- Ammonium Hydroxide Precipitate:≤0.02 %
- Sodium (Na):≤0.05 %
- Nitrogen Compounds (as N):≤10 ppm
- Phosphate (PO₄):≤5 ppm
- Heavy Metals (as Ag):≤10 ppm
- Iron (Fe):≤10 ppm
- Mercury (Hg), For Information Only:
- Nickel (Ni):≤10 ppm
- Calcium (Ca):≤50 ppm
- Magnesium (Mg), For Information Only:
- J.T.Baker®, Potassium hydroxide ≥85.0%, pellets, BAKER ANALYZED®, Electronic Grade:
Specification Test Results
Related Information
Certificates
Search by Lot Number3141 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Brand Definition Baker Analyzed
Datasheet
sds_BAKR3141
SDS
Customers Who Bought This Also Bought