Potassium citrate monohydrate ≥99%, granular FCC, USP, J.T.Baker®
Supplier: Avantor
MOC: Regulated
About this item
Synonyms: Potassium citrate tribasic monohydrate,Citric acid tripotassium salt monohydrate,Tripotassium citrate monohydrate,Tripotassium 2-hydroxypropane-1,2
Formula:HOC(COOK)(CH₂COOK)₂·H₂O
MW:324.41 g/mol
Melting Pt:230 °C
Density:1.98 g/cm³ (20 °C)
MDL Number:MFCD00150442
CAS Number:6100-05-6
Merck Index:13,07706
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 230 °C
:Flash Pt:
:Flash Pt:
:M.W. 324.41 g/mol
:Potassium citrate monohydrate
EINECS:212-755-5
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- GMP Manufactured Product:
- Food GMP Manufactured Product:
- Meets U.S.P Requirements:
- Meets F.C.C. Requirements:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Food Chemical:
- Bulk Pharmaceutical Chemical:
- USP - Identification:Passes Test
- USP - Alkalinity:Passes Test
- USP - Loss on Drying:3.0 - 6.0 %
- USP - Tartrate:Passes Test
- USP - Assay (C₆H₅K₃O₇) (dried basis):99.0 - 100.5 %
- FCC - Identification:Passes Test
- FCC - Alkalinity:Passes Test
- FCC - Lead (Pb):≤2 mg/kg
- FCC - Loss on Drying:3.0 - 6.0 %
- FCC - Assay (C₆H₅K₃O₇) (dried basis):99.0 - 100.5 %
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Metallic Residues: No metal catalysts or metal reagents, as defined by:
- EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of:
- this material.:
- J.T.Baker®, Potassium citrate monohydrate ≥99%, granular FCC, USP:
Specification Test Results
Related Information
Certificates
Search by Lot Number3068 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Customers Who Bought This Also Bought