Potassium citrate monohydrate 99.0-101.0%, granular, BAKER ANALYZED®, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
Synonyms: Potassium citrate tribasic monohydrate,Citric acid tripotassium salt monohydrate,Tripotassium citrate monohydrate,Tripotassium 2-hydroxypropane-1,2
Formula:HOC(COOK)(CH₂COOK)₂·H₂O
MW:324.41 g/mol
Melting Pt:230 °C
Density:1.98 g/cm³ (20 °C)
MDL Number:MFCD00150442
CAS Number:6100-05-6
Merck Index:13,07706
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-101,0
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 230 °C
:Flash Pt:
:Flash Pt:
:M.W. 324.41 g/mol
:Potassium citrate monohydrate
EINECS:212-755-5
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Assay (HOC(COOK)(CH₂COOK)₂·H₂O):99.0 - 101.0 %
- Insoluble Matter:≤0.005 %
- pH of 5% Solution at 25°C:7.0 - 9.0
- Chloride (Cl):≤0.001 %
- Sulfate (SO₄):≤0.005 %
- Calcium (Ca):≤0.005 %
- Trace Impurities - Iron (Fe):≤0.001 %
- Sodium (Na):≤0.2 %
- Phosphate (PO₄):≤5 ppm
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number3066 Product Specifications
Additional Product Details
Brand Definition Baker Analyzed
Datasheet
sds_BAKR3066
SDS
Customers Who Bought This Also Bought