Sodium phosphate dibasic, anhydrous ≥99.0%, powder, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated, With Information
About this item
Synonyms: di-Sodium hydrogen orthophosphate,Sodium hydrogen phosphate
Formula:Na₂HPO₄
MW:141.96 g/mol
Melting Pt:250 °C
Density:1.53 g/cm³ (20 °C)
MDL Number:MFCD00003496
CAS Number:7558-79-4
Merck Index:12,08805
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0
:Management of Change (MOC) category = NI
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 250 °C
:Flash Pt:
:Flash Pt:
:M.W. 141.96 g/mol
:Sodium phosphate dibasic
Storage Temperature:Ambient
EINECS:231-448-7
REACH:01-2119489797-11
:M.O.C. Category NI
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Assay (Na₂HPO₄):≥99.0 %
- DNase Activity (None Detected):Passes Test
- RNase Activity:Passes Test
- Protease Activity:Passes Test
- pH of 5% Solution at 25°C:8.7 - 9.3
- Insoluble Matter:≤0.01 %
- Heavy Metals (as Pb):≤0.001 %
- Trace Impurities - Iron (Fe):≤0.001 %
- Loss on Drying at 105°C:≤0.2 %
Specification Test Results
Related Information
Certificates
Search by Lot Number4062 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Customers Who Bought This Also Bought