Sodium phosphate dibasic, anhydrous ≥98% USP, ACS endotoxin tested, J.T.Baker®
Catalog #: CAJT3826-7
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 7558-79-4
MOC: Highly RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:PE pail, screw top
- Size:12 kg
- Cat. no.:CAJT3826-7
- Supplier No.:3826-07
Specifications
Specification Test Results
- Meets U.S.P Requirements:
- GMP Manufactured Product:
- Meets ACS Reagent Chemical Requirements:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Assay (Na₂HPO₄) (dried basis):98.0 - 100.5 %
- USP - Arsenic (As):≤ 3 ppm
- USP - Chloride (Cl):≤ 0.06 %
- USP - Identification:Passes Test
- USP - Insoluble Matter:≤ 0.2 %
- USP - Loss on Drying at 130°C:≤ 5.0 %
- USP - Sulfate (SO₄):≤ 0.2 %
- ACS - Assay (Na₂HPO₄) (by acidimetry):≥ 99.0 %
- ACS - Chloride (Cl):≤ 0.002 %
- ACS - Heavy Metals (as Pb):≤ 0.001 %
- ACS - Insoluble Matter:≤ 0.01 %
- ACS - Iron (Fe):≤ 0.001 %
- ACS - Loss on Drying at 105°C:≤ 0.2 %
- ACS - pH of 5% Solution at 25°C:8.7 - 9.3
- ACS - Sulfate (SO₄):≤ 0.005 %
- Endotoxin Concentration (5 EU/g max.):Passes Test
- (Suitable for Buffer Solutions):
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- J.T.Baker®, Sodium phosphate dibasic, anhydrous ≥98% USP, ACS endotoxin tested:
Specification Test Results
About this item
Synonyms: sec-Sodium phosphate,di-Sodium hydrogen orthophosphate,di-Sodium hydrogen phosphate anhydrous,Sodium hydrogen phosphate,Disodium phosphate
Melting Pt:250 °C
Density:1.53 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98
:Management of Change (MOC) category = HR
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 250 °C
:Flash Pt:
:Flash Pt:
:M.W. 141.96 g/mol
Merck Index:12,08805
:Sodium phosphate dibasic
Formula:Na₂HPO₄
MW:141.96 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00003496
CAS Number:7558-79-4
EINECS:231-448-7
REACH:01-2119489797-11
:M.O.C. Category HR
Customers Who Bought This Also Bought