Disodium hydrogen phosphate heptahydrate 98-100.0% (dried basis), crystals USP, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: Sodium phosphate dibasic heptahydrate,Sodium hydrogen phosphate heptahydrate
Formula:HNa₂O₄P·7H₂O
MW:268.07 g/mol
Melting Pt:48 °C
Density:1.7 g/cm³ (20 °C)
MDL Number:MFCD00149180
CAS Number:7782-85-6
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98-100,0
:Management of Change (MOC) category = HR
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 48 °C
:Flash Pt:
:Flash Pt:
:M.W. 268.07 g/mol
Storage Temperature:Ambient
EINECS:231-448-7
:M.O.C. Category HR
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- GMP Manufactured Product:
- Meets U.S.P Requirements:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Assay (Na₂HPO₄) (dried basis):98.0 - 100.5 %
- USP - Arsenic (As):≤16 ppm
- USP - Chloride (Cl):≤0.06 %
- USP - Identification:Passes Test
- USP - Insoluble Substances:≤0.4 %
- USP - Loss on Drying at 130°C:43.0 - 50.0 %
- USP - Sulfate (SO₄):≤0.2 %
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
Specification Test Results
Related Information
Certificates
Search by Lot Number3817 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals VWR Top Rated Guide
Product Literature
spec_JT3817-5 spec_JT3817-7
Product specification
Customers Who Bought This Also Bought