Sodium phosphate dibasic, anhydrous 98.0-100.5% (dried basis) USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
Synonyms: sec-Sodium phosphate,di-Sodium hydrogen orthophosphate,di-Sodium hydrogen phosphate anhydrous,Sodium hydrogen phosphate,Disodium phosphate
Formula:Na₂HPO₄
MW:141.96 g/mol
Melting Pt:250 °C
Density:2.07 g/cm³ (20 °C)
MDL Number:MFCD00003496
CAS Number:7558-79-4
Merck Index:12,08805
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,0-100,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 250 °C
:Flash Pt:
:Flash Pt:
:M.W. 141.96 g/mol
:Sodium phosphate dibasic
Storage Temperature:Ambient
EINECS:231-448-7
REACH:01-2119489797-11
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets JPE Specifications:
- Meets E.P. Chemical Specifications:
- Meets U.S.P Requirements:
- GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Assay (Na₂HPO₄) (dried basis):98.0 - 100.5 %
- USP - Identification:Passes Test
- USP - Loss on Drying at 130°C:≤ 5.0 %
- USP - Insoluble Substances:≤ 0.4 %
- USP - Chloride (Cl):≤ 0.06 %
- USP - Sulfate (SO₄):≤ 0.2 %
- USP - Arsenic (As):≤ 16 ppm
- USP - Endotoxin Concentration (2.5 EU/g max.):Passes Test
- JPE - Assay (dried basis):≥ 99.0 %
- JPE - Identification:Passes Test
- JPE - Arsenic (As):≤ 2 ppm
- JPE - Carbonate:Passes Test
- JPE - Chloride (Cl):≤ 0.011 %
- JPE - Clarity and Color of Solution:Passes Test
- JPE - Heavy Metals:≤ 10 ppm
- JPE - pH (1-100):9.0 - 9.4
- JPE - Sulfate (SO₄):≤ 0.038 %
- JPE - Loss on Drying:≤ 2.0 %
- EP - Assay (Na₂HPO₄) (dried basis):98.0 - 101.0 %
- EP - Appearance of Solution:Passes Test
- EP - Identification A:Passes Test
- EP - Identification B:Passes Test
- EP - Identification C:Passes Test
- EP - Identification D:Passes Test
- EP - Reducing Substances:Passes Test
- EP - Chloride (Cl):≤ 200 ppm
- EP - Iron (Fe):≤ 20 ppm
- EP - Loss on Drying:≤ 1.0 %
- EP - Monosodium Phosphate:≤ 2.5 %
- EP - Sulfate (SO₄):≤ 500 ppm
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
Specification Test Results
Related Information
Certificates
Search by Lot Number3804 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals VWR Top Rated Guide
Product Literature
sds_BAKR3804
SDS
spec_JT3804-08 spec_JT3804-1 spec_JT3804-65
Product specification
Customers Who Bought This Also Bought