Sodium hydroxide 95.0-100.5%, pellets FCC, NF, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: Caustic soda
Formula:NaOH
MW:40 g/mol
Boiling Pt:1390 °C (1013 hPa)
Melting Pt:323 °C
Density:2.13 g/cm³ (20 °C)
MDL Number:MFCD00003548
CAS Number:1310-73-2
ADR:8,II
Merck Index:14,08627
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95,0-100,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 1390 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 323 °C
:Flash Pt:
:Flash Pt:
:M.W. 40 g/mol
EINECS:215-185-5
UN:1823
REACH:01-2119457892-27
:8
:II
:GHS05
Storage Temperature:Ambient
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Food GMP Manufactured Product:
- GMP Manufactured Product:
- Meets N.F. Requirements:
- Meets F.C.C. Requirements:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Food Chemical:
- Bulk Pharmaceutical Chemical:
- NF - Identification A (Sodium):Passes Test
- NF - Identification B (pH):≥11.0
- NF - Insoluble Substances and Organic Matter:Passes Test
- NF - Potassium (K):≤0.5 %
- NF - Total Alkali:95.0 - 100.5 %
- NF - Content of Sodium:54.0 - 59.8 %
- NF - Carbonate (as Na₂CO₃):≤3.0 %
- FCC - Insoluble Substances and Organic Matter:Passes Test
- FCC - Assay:95.0 - 100.5 %
- FCC - Carbonate (as Na₂CO₃):≤3.0 %
- FCC - Arsenic (As):≤3 ppm
- FCC - Lead (Pb):≤2 mg/kg
- FCC - Mercury (Hg):≤0.1 ppm
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- STORAGE: Preserve in tight containers.:
- Elemental Impurities (USP 232) - Information on elemental impurities for:
- this product is available on the associated Product Regulatory Data:
- Sheet and elemental impurity profile report.:
Specification Test Results
Related Information
Certificates
Search by Lot Number3728 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
JTBaker_3728_Datasheet.pdf
Datasheet
spec_JT3728-20 spec_JT3728-7 spec_JT3728-70
Product specification
Customers Who Bought This Also Bought