Sodium hydroxide solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®
Supplier: VWR International
MOC: Non-Regulated
About this item
Synonyms: Caustic soda
Formula:NaOH
MW:40 g/mol
Boiling Pt:119…135 °C (1013 hPa)
Melting Pt:9…10 °C
Density:1.53 g/cm³ (20 °C)
MDL Number:MFCD00003548
CAS Number:1310-73-2
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt: 119…135 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 323 °C|Melting Pt: 9…10 °C
:Flash Pt:
:Flash Pt:
:M.W. 40 g/mol
Storage Temperature:Ambient
EINECS:215-185-5
:119…135 °C (1013 hPa)
UN:1824
:8
:II
:GHS05
:GHS07
:M.O.C. Category N
:12,08772
:14,08627
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Protect from air to avoid absorption of carbon dioxide.:
- Assay (NaOH) (by acidimetry):50 - 52 %
- Color (APHA):≤20
- Sodium Carbonate (Na₂CO₃):≤0.1 %
- Chloride (Cl):≤0.002 %
- Sulfate (SO₄):≤0.001 %
- Ammonium Hydroxide Precipitate:≤0.01 %
- Silica (SiO₂), For Information Only:
- Heavy Metals (as Ag):≤0.001 %
- Potassium (K):≤0.01 %
- Nitrogen Compounds (as N):≤5 ppm
- Phosphate (PO₄):≤5 ppm
- Trace Impurities - Iron (Fe):≤5 ppm
- Trace Impurities - Mercury (Hg), For Information Only:
- Trace Impurities - Nickel (Ni):≤5.000 ppm
- IMPORTANT: Material will freeze if stored below 60°F (16°C).:
Specification Test Results
Related Information
Certificates
Search by Lot Number3727 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor High-Purity Acids Avantor Portfolio of Chemicals Avantor™ High-Purity Acids VWR Top Rated Guide
Product Literature
spec_JT3727-3 spec_JT3727-9
Product specification
Customers Who Bought This Also Bought