AlphaTec® 19-026 Chemical and heat resistant gloves, Ansell
Supplier: Ansell Healthcare
Certificates
About this item
Type 19-026 neoprene gloves feature a comfortable, heat-resistant thermal liner.
- Hand-specific
- Smooth finish
- Straight cuffs
Flexible gloves combine improved dexterity with protection from hot surfaces and chemical splash. Gloves provide protection from intermittent contact with hot surfaces up to 204 °C (400 °F).
Specifications
- Catalog No:
- 47742-582
- 89009-080
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific