Sodium acetate, anhydrous ≥99% FCC, USP endotoxin tested, J.T.Baker®
Catalog #: CAJT3474-7
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 127-09-3
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:PE pail, screw top
- Size:12 kg
- Cat. no.:CAJT3474-7
- Supplier No.:3474-07
Specifications
Specification Test Results
- GMP Manufactured Product:
- Meets F.C.C. Requirements:
- Meets U.S.P Requirements:
- Food GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Food Chemical:
- Bulk Pharmaceutical Chemical:
- USP - Assay (CH₃COONa) (dried basis):99.0 - 101.0 %
- USP - Calcium and Magnesium:Passes Test
- USP - Chloride (Cl):≤0.035 %
- USP - Identification:Passes Test
- USP - Insoluble Matter:≤0.05 %
- USP - Loss on Drying at 120°C:≤1.0 %
- USP - pH of 3% Solution at 25°C:7.5 - 9.2
- USP - Potassium Compounds:Passes Test
- USP - Residual Acetic Acid, For Information Only:
- USP - Sulfate (SO₄):≤0.005 %
- FCC - Assay (CH₃COONa) (dried basis):99.0 - 101.0 %
- FCC - Alkalinity:≤0.2 %
- FCC - Identification:Passes Test
- FCC - Lead (Pb):≤2 mg/kg
- FCC - Loss on Drying at 120°C:≤1.0 %
- FCC - Potassium Compounds:Passes Test
- Endotoxin Concentration (2.5 EU/g max.):Passes Test
- The Class 3 solvent, acetic acid, is used to produce this product.:
- Residual concentration is controlled to <0.5%.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- J.T.Baker®, Sodium acetate, anhydrous ≥99% FCC, USP endotoxin tested:
Specification Test Results
About this item
Synonyms: Acetic acid sodium salt
Boiling Pt:881.4 °C (1013 hPa)
Melting Pt:324 °C
Density:1.528 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99
:Management of Change (MOC) category = R
Flash Pt:>250 °C (closed cup)
:Boiling Pt:
:Boiling Pt: 881.4 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 324 °C
:Flash Pt:
:Flash Pt: >250 °C (closed cup)
:M.W. 82.03 g/mol
Merck Index:12,08711
Formula:H₃CCOONa
MW:82.03 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00012459
CAS Number:127-09-3
EINECS:204-823-8
REACH:01-2119485123-42
:>250 °C
:M.O.C. Category R
Customers Who Bought This Also Bought