Sodium acetate, anhydrous ≥99% FCC, USP, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: Acetic acid sodium salt
Formula:H₃CCOONa
MW:82.03 g/mol
Boiling Pt:881.4 °C (1013 hPa)
Melting Pt:324 °C
Density:1.528 g/cm³ (20 °C)
Flash Pt:>250 °C (closed cup)
MDL Number:MFCD00012459
CAS Number:127-09-3
Merck Index:12,08711
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 881.4 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 324 °C
:Flash Pt:
:Flash Pt: >250 °C (closed cup)
:M.W. 82.03 g/mol
Storage Temperature:Ambient
EINECS:204-823-8
REACH:01-2119485123-42
:>250 °C
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Food GMP Manufactured Product:
- GMP Manufactured Product:
- Meets F.C.C. Requirements:
- Meets U.S.P Requirements:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- Bulk Food Chemical:
- USP - Assay (CH₃COONa) (dried basis):99.0 - 101.0 %
- USP - Calcium and Magnesium:Passes Test
- USP - Chloride (Cl):≤0.035 %
- USP - Identification:Passes Test
- USP - Insoluble Matter:≤0.05 %
- USP - Loss on Drying at 120°C:≤1.0 %
- USP - pH of 3% Solution at 25°C:7.5 - 9.2
- USP - Potassium Compounds:Passes Test
- USP - Residual Acetic Acid, For Information Only:
- USP - Sulfate (SO₄):≤0.005 %
- FCC - Assay (CH₃COONa) (dried basis):99.0 - 101.0 %
- FCC - Alkalinity:≤0.2 %
- FCC - Identification:Passes Test
- FCC - Lead (Pb):≤2 mg/kg
- FCC - Loss on Drying at 120°C:≤1.0 %
- FCC - Potassium Compounds:Passes Test
- The Class 3 solvent, acetic acid, is used to produce this product.:
- Residual concentration is controlled to <0.5%.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- J.T.Baker®, Sodium acetate, anhydrous ≥99% FCC, USP:
Specification Test Results
Related Information
Certificates
Search by Lot Number3473 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals VWR Top Rated Guide
Product Literature
spec_JT3473-5 spec_JT3473-QM spec_JT3473-R
Product specification
Customers Who Bought This Also Bought