Sodium acetate trihydrate 99.0-101.0% (dried basis), crystals USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Highly Regulated
About this item
Synonyms: Acetic acid, sodium salt, trihydrate,Plasmafusin
Formula:H₃CCOONa·3H₂O
MW:136.08 g/mol
Boiling Pt:120…123 °C (1013 hPa)
Melting Pt:58 °C
Density:1.45 g/cm³
MDL Number:MFCD00071557
CAS Number:6131-90-4
Merck Index:12,08711
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-101,0
:Management of Change (MOC) category = HR
:Boiling Pt:
:Boiling Pt: 120…123 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 58 °C
:Flash Pt:
:Flash Pt:
:M.W. 136.08 g/mol
EINECS:204-823-8
:120…123 °C (1013 hPa)
Storage Temperature:Ambient
:M.O.C. Category HR
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets U.S.P Requirements:
- GMP Manufactured Product:
- Meets F.C.C. Requirements:
- Food GMP Manufactured Product:
- Meets J.P. Chemical Specifications:
- Meets E.P. Chemical Specifications:
- Meets B.P. Chemical Specifications:
- Preserve in tight containers, store at room temperature.:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Food Chemical:
- Bulk Pharmaceutical Chemical:
- USP - Assay (CH₃COONa) (dried basis):99.0 - 101.0 %
- USP - Calcium and Magnesium:Passes Test
- USP - Chloride (Cl):≤ 0.035 %
- USP - Endotoxin Concentration (EU/g):≤ 2.5
- USP - Identification:Passes Test
- USP - Insoluble Matter:≤ 0.05 %
- USP - Loss on Drying:38.0 - 41.0 %
- USP - pH of 5% Solution at 25°C:7.5 - 9.2
- USP - Potassium (K):Passes Test
- USP - Sulfate (SO₄):≤ 0.005 %
- EP/BP - Assay (CH₃COONa):99.0 - 101.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Identification C:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - pH (1 in 20):7.5 - 9.0
- EP/BP - Reducing Substances:Passes Test
- EP/BP - Chloride (Cl):≤ 200 ppm
- EP/BP - Sulfate (SO₄):≤ 200 ppm
- EP/BP - Calcium and Magnesium:≤ 50 ppm
- EP/BP - Iron (Fe):≤ 10 ppm
- EP/BP - Loss on Drying:39.0 - 40.5 %
- JP - Assay (CH₃COONa):99.5 - 101.0 %
- JP - Identification:Passes Test
- JP - Clarity and Color of Solution:Passes Test
- JP - Acidity or Alkalinity:Passes Test
- JP - Chloride (Cl):≤ 0.011 %
- JP - Sulfate (SO₄):≤ 0.017 %
- JP - Heavy Metals (as Pb):≤ 10 ppm
- JP - Calcium and Magnesium:Passes Test
- JP - Arsenic (As):≤ 2 ppm
- JP - Substances Reducing Permanganate:Passes Test
- JP - Loss on Drying:39.0 - 40.5 %
- FCC - Alkalinity:≤ 0.05 %
- FCC - Assay (CH₃COONa) (dried basis):99.0 - 101.0 %
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- FCC - Lead (Pb):≤ 2 mg/kg
- FCC - Loss on Drying:38.0 - 41.0 %
- FCC - Potassium Compounds:Passes Test
- No Class 1, 2, 3 or other solvent is likely to be present in the final:
- product.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- J.T.Baker®, Sodium acetate trihydrate 99.0-101.0% (dried basis), crystals USP, Multi-Compendial:
Specification Test Results
Related Information
Certificates
Search by Lot Number3461 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals VWR Top Rated Guide
Product Literature
spec_JT3461-06 spec_JT3461-1 spec_JT3461-10 spec_JT3461-19 spec_JT3461-5
Product specification
More
Customers Who Bought This Also Bought