Order Entry
Canada
ContactUsLinkComponent
Silica gel, BAKER ANALYZED® 80-200 Mesh, J.T.Baker®
Silica gel, BAKER ANALYZED® 80-200 Mesh, J.T.Baker®
Catalog # CAJT3404-1
CAS Number:  
MOC:  Non-RegulatedCertificates
undefined
Silica gel, BAKER ANALYZED® 80-200 Mesh, J.T.Baker®
Catalog # CAJT3404-1
Supplier Number:  BAKR3404-01
CAS Number:  
MOC:  Non-RegulatedCertificates

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle, amber, wide mouth
  • Size:
    500 g
  • Cat. no.:
    CAJT3404-1
  • Supplier No.:
    3404-01

Specifications

Specification Test Results

  • Loss on Ignition:
    ≤13 %
  • Alcohol-Ether Soluble Substances:
    ≤0.05 %
  • Flow Rate, mL/min.:
    ≥0.5
  • Mesh - Thru U.S. No. 80 Sieve:
    ≥90.0 %
  • Mesh - Thru U.S. No. 200 Sieve:
    ≤10.0 %

Specification Test Results

About this item

 :
 :60.08 g/mol
Boiling Pt:2230 °C (1013 hPa)
Melting Pt:1710 °C
Density:2.10 kg/m³
 :Management of Change (MOC) category = N
Storage Temperature:Ambient
REACH:01-2119379499-16
CAS Number:63231-67-4
 :7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232