Parteck® SI 400 LEX (Sorbitol), EMPROVE® exp Ph. Eur., BP, NF, JP suitable for use as excipient, SAFC®
Supplier: EMD MILLIPORE – PCS CA EXCEP
Certificates
About this item
Synonyms: (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol,(Sorbitol),D(-)-Glucitol
CAS Number:50-70-4
:98201-93-5
Formula:HOH₂C(CH(OH))₄CH₂OH
MW:182.17 g/mol
Boiling Pt:295 °C (1013 hPa)
Melting Pt:94…98 °C
Density:1.524 g/cm³ (20 °C)
Flash Pt:>100 °C
MDL Number:MFCD00004708
Merck Index:12,08873
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt: 295 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 94…98 °C
:Flash Pt:
:Flash Pt: >100 °C
:M.W. 182.17 g/mol
:Parteck® SI 400 LEX (Sorbitol)
EINECS:200-061-5
:>100 °C
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Assay (HPLC, calc.on anhydrous substance):97.0 - 100.5 %
- Identity:passes test
- pH-value (10 %; water):3.5 - 7.0
- Appearance of solution (25 %; water):passes test
- Spec. rotation (α20/D, 10 %, borate complex, calc. on anhydrous substance):+4.0 - +7.0 °
- Conductivity (25°C) (20 %; water):≤ 20
- Chloride (Cl):≤ 0.002 %
- Sulfate (SO₄):≤ 0.006 %
- Heavy metals (as Pb):≤ 5 ppm
- As (Arsenic):≤ 1.3 ppm
- Ni (Nickel) (#):≤ 1 ppm
- Pb (Lead):≤ 0.5 ppm
- Related substances (HPLC, EP) (Major impurity (Mannitol)):≤ 2.0 %
- Related substances (HPLC, EP) (Sum of all impurities):≤ 3.0 %
- Reducing sugars after hydrolysis/total sugar (as glucose):≤ 0.8
- Reducing sugars (as glucose):≤ 0.11 %
- Sulfated ash:≤ 0.02 %
- Loss on drying:≤ 2.0 %
- Water:≤ 1.5 %
- Residual solvents (ICH Q3C):excluded by manufacturing process
- Microbiological test:passes test
- Colony count (aerobic bacteria (TAMC)):≤ 10²
- Colony count (Yeasts and moulds (TYMC)):≤ 10²
- Salmonella spp.:in 10g Substance not detectable
- E.coli:in 1g substance not detectable
- Pseudomonas aeruginosa:in 1g substance not detectable
- Staphylococcus aureus:in 1g substance not detectable
- Candida albicans:In 1g substance not detectable
- Endotoxins:≤ 1
Specification Test Results
Customers Who Bought This Also Bought