Chem Master Neoprene/Rubber Gloves, Showa
Supplier: Showa-Best Glove
Certificates
About this item
Hand and product protection for manufacturing, electronics, machine work, food processing, pharmaceutical, and laboratory applications.
- Resists a broad range of chemicals in many applications
- 100% cotton flock lining
- Provide comfort, flexibility, and tactility
- Self-flushing tractor tread grip for fluid runoff increases gripping effectiveness
Unsupported neoprene over natural rubber construction offers abrasion, tear, and puncture resistantance.
Color: black over blue.
Certifications: Sanitized gloves are made using only listed materials and levels outlined in 21 CFR 170-199.
Specifications
- Catalog No:
- CA32892-058
- CA32892-057
- CA32892-060
- CA32892-059
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Finish:
- Tractor Tread
- Tractor Tread
- Tractor Tread
- Tractor Tread
- Cuff style:
- Straight
- Straight
- Straight
- Straight