Potassium acetate 99.0-100.5% (dry basis), granular USP, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
Synonyms: Acetic acid potassium salt
Formula:CH₃COOK
MW:98.14 g/mol
Melting Pt:304 °C
Density:1.57 g/cm³ (25 °C)
MDL Number:MFCD00012458
CAS Number:127-08-2
Merck Index:12,07764
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-100,5
:Management of Change (MOC) category = R
Flash Pt:>250 °C
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 304 °C
:Flash Pt:
:Flash Pt: >250 °C
:M.W. 98.14 g/mol
EINECS:204-822-2
:>250 °C
Storage Temperature:Ambient
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- GMP Manufactured Product:
- Meets U.S.P Requirements:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Assay (CH₃COOK) (dried basis):99.0 - 100.5 %
- USP - Identification:Passes Test
- USP - Loss on Drying:≤1.0 %
- USP - pH (1 in 20):7.5 - 8.5
- USP - Residual Acetic Acid, For Information Only:
- USP - Sodium (Na):≤0.03 %
- Only Class 3 Solvents are likely to be present. Residual concentration of all Class 3 Solvents is <0.5%.:
- J.T.Baker®, Potassium acetate 99.0-100.5% (dry basis), granular USP:
Specification Test Results
Related Information
Certificates
Search by Lot Number2914 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
sds_BAKR2914
SDS
spec_JT2914-R
Product specification
Customers Who Bought This Also Bought